Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trihydroxybenzoic Acid Monohydrate 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
71989-93-0
C7H8O6
188.135
MFCD00149095
HWZIRFCGHAROOI-UHFFFAOYSA-N
2,4,6-trihydroxybenzoic acid hydrate,2,4,6-trihydroxybenzoic acid monohydrate,benzoic acid, 2,4,6-trihydroxy-, monohydrate,phloroglucinolcarboxylic acid,pubchem20337,acmc-209olb,2,4,6-trihydroxybenzoicacidmonohydrate,2,4,6-trihydroxybenzoic acid, hydrate,2,4,6-trihydroxybenzoic acid monohydrate, technical t,2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
2723793
2,4,6-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)C(=O)O)O)O.O
71989-93-0
C7H8O6
188.135
MFCD00149095
HWZIRFCGHAROOI-UHFFFAOYSA-N
2,4,6-trihydroxybenzoic acid hydrate,2,4,6-trihydroxybenzoic acid monohydrate,benzoic acid, 2,4,6-trihydroxy-, monohydrate,phloroglucinolcarboxylic acid,pubchem20337,acmc-209olb,2,4,6-trihydroxybenzoicacidmonohydrate,2,4,6-trihydroxybenzoic acid, hydrate,2,4,6-trihydroxybenzoic acid monohydrate, technical t,2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
2723793
2,4,6-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)C(=O)O)O)O.O
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Color Beige
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (HO)3C6H2CO2H·H2O
Formula Weight 188.14
Physical Form Micro-Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material 2, 4, 6-Trihydroxybenzoic acid monohydrate, 95%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-467-5

RTECSNumber : DH8910000

TSCA : TSCA