Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triiodophenol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
609-23-4
C6H3I3O
471.802
MFCD00002179
VAPDZNUFNKUROY-UHFFFAOYSA-N
phenol, 2,4,6-triiodo,triiodophenol,unii-9rb2r81a7u,2,4,6-trijodfenol,2,4,6-trijodfenol czech,2,4,6-triiodo-phenol,chembl81721,2,4,6-triiodo-phenol tip,2,6-triiodophenol,phenol,4,6-triiodo
11862
2,4,6-triiodophenol
C1=C(C=C(C(=C1I)O)I)I
609-23-4
C6H3I3O
471.802
MFCD00002179
VAPDZNUFNKUROY-UHFFFAOYSA-N
phenol, 2,4,6-triiodo,triiodophenol,unii-9rb2r81a7u,2,4,6-trijodfenol,2,4,6-trijodfenol czech,2,4,6-triiodo-phenol,chembl81721,2,4,6-triiodo-phenol tip,2,6-triiodophenol,phenol,4,6-triiodo
11862
2,4,6-triiodophenol
C1=C(C=C(C(=C1I)O)I)I
Color White
Melting Point 160°C
Quantity 25g
Formula Weight 471.80
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 2,4,6-Triiodophenol

RTECSNumber : SN2800000

TSCA : Yes