Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triisopropylbenzenesulfonyl azide, 97%, contains 15% water as stabilizer, ACROS Organics™

Click to view available options
Quantity:
1g
5g
36982-84-0
C15H23N3O2S
309.43
AEMWUHCKKDPRSK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl azide,2,4,6-triisopropylbenzene-sulfonyl azide,trisyl azide,2,4,6-triisopropylbenzene-sulfonyl azide,,benzenesulfonyl azide, 2,4,6-tris 1-methylethyl,2,4,6-triisopropyl-benzenesulfonyl azide,n-diazo-2,4,6-triisopropyl-benzenesulfonamide,2,4,6-tris 1-methylethyl-benzenesulfonyl azide,2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water,trisyl azide solution
371707
N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C
36982-84-0
C15H23N3O2S
309.43
AEMWUHCKKDPRSK-UHFFFAOYSA-N
2,4,6-triisopropylbenzenesulfonyl azide,2,4,6-triisopropylbenzene-sulfonyl azide,trisyl azide,2,4,6-triisopropylbenzene-sulfonyl azide,,benzenesulfonyl azide, 2,4,6-tris 1-methylethyl,2,4,6-triisopropyl-benzenesulfonyl azide,n-diazo-2,4,6-triisopropyl-benzenesulfonamide,2,4,6-tris 1-methylethyl-benzenesulfonyl azide,2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water,trisyl azide solution
371707
N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C
Melting Point 41.0°C to 43.0°C
Packaging Glass Bottle
Quantity 1g
Water 10 to 20%
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 309.43
Percent Purity 97%
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl azide

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash

GHS Signal Word: Danger