Alfa Aesar™ 2,4,6-Triisopropylbenzenesulfonyl chloride, 98%

Manufacturer: Alfa Aesar™ A1145822

Quantity 100g

 View more versions of this product

Catalog No. AAA1145822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6553-96-4
    Molecular Formula C15H23ClO2S
    Formula Weight 302.87
    MDL Number MFCD00007433
    UN Number UN3261
    Beilstein 1218575
    Synonym Trisyl chloride
    Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl chloride

    Specifications

    Formula Weight 302.87
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 6553-96-4
    UN Number UN3261
    Synonym Trisyl chloride
    Quantity 100g
    Molecular Formula C15H23ClO2S
    Beilstein 1218575
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    MDL Number MFCD00007433
    Health Hazard 1 Danger
    Melting Point 94° to 97°C
    Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl chloride
    Assay 98%

    Description

    Applications
    2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.

    Solubility
    Soluble in chloroform.

    Notes
    Incompatible with strong bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit