Alfa Aesar™ 2,4,6-Triisopropylbenzoic acid, 97%

Manufacturer: Alfa Aesar™ A1548909

Quantity 10g

 View more versions of this product

Catalog No. AAA1548909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 49623-71-4
    Molecular Formula C16H24O2
    Formula Weight 248.37
    MDL Number MFCD00015031
    Beilstein 2372331
    Chemical Name or Material 2,4,6-Triisopropylbenzoic acid

    Specifications

    Formula Weight 248.37
    Molecular Formula C16H24O2
    MDL Number MFCD00015031
    CAS 49623-71-4
    Beilstein 2372331
    Melting Point 184° to 187°C
    Quantity 10g
    Chemical Name or Material 2,4,6-Triisopropylbenzoic acid
    Assay 97%

    Description

    Applications
    2,4,6-Triisopropylbenzoic acid is a hindered acid intermediate compound in organic synthesis.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit