Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
250g
Packaging:
Glass bottle
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy,2,4,6-trimethoxybenz aldehyde,pubchem8264,acmc-1cuhn,ksc490m2h,2,4,6-trimethoxy-benzaldehyde,2,4,6-trimethoxybenz-aldehyde,2,4,6-trimethoxy benz aldehyde,crbzvdlxaiferf-uhfffaoysa,1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC
830-79-5
C10H12O4
196.2
MFCD00003313
CRBZVDLXAIFERF-UHFFFAOYSA-N
benzaldehyde, 2,4,6-trimethoxy,2,4,6-trimethoxybenz aldehyde,pubchem8264,acmc-1cuhn,ksc490m2h,2,4,6-trimethoxy-benzaldehyde,2,4,6-trimethoxybenz-aldehyde,2,4,6-trimethoxy benz aldehyde,crbzvdlxaiferf-uhfffaoysa,1-formyl-2,4,6-trimethoxybenzene
70019
2,4,6-trimethoxybenzaldehyde
COC1=CC(=C(C(=C1)OC)C=O)OC
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 250g
Melting Point 118.0°C to 120.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)3C6H2CHO
Beilstein 08,39
Formula Weight 196.2
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trimethoxybenzaldehyde, 98%
Show More Show Less

EINECSNumber : 212-598-2

RTECSNumber : CU8460200

TSCA : TSCA