Alfa Aesar™ 2,4,6-Trimethoxybenzonitrile, 98%

Manufacturer: Alfa Aesar™ B2354003

Quantity 1g

 View more versions of this product

Catalog No. AAB2354003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2571-54-2
    Molecular Formula C10H11NO3
    Formula Weight 193.2
    MDL Number MFCD00001787
    Chemical Name or Material 2,4,6-Trimethoxybenzonitrile

    Specifications

    Formula Weight 193.2
    Molecular Formula C10H11NO3
    MDL Number MFCD00001787
    CAS 2571-54-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P280h
    Wear protective gloves/clothing.
    Odor Characteristic
    Melting Point 140° to 143°C
    Quantity 1g
    Chemical Name or Material 2,4,6-Trimethoxybenzonitrile
    Assay 98%

    Description

    Applications
    2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280h
    Wear protective gloves/clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit