Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylphenylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
5980-97-2
C9H13BO2
164.01
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
292184
(2,4,6-trimethylphenyl)boronic acid
B(C1=C(C=C(C=C1C)C)C)(O)O
5980-97-2
C9H13BO2
164.01
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
292184
(2,4,6-trimethylphenyl)boronic acid
B(C1=C(C=C(C=C1C)C)C)(O)O
Quantity 25g
Melting Point 131.0°C to 136.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Formula Weight 164.01
Percent Purity 95%
Chemical Name or Material 2,4,6-Trimethylphenylboronic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning