Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,6-Trimethylthiophenol, 97%

Click to view available options
Quantity:
1g
5g
25g
1541-10-2
C9H12S
152.255
MFCD00218475
PKANQZUPJCMBAK-UHFFFAOYSA-N
2,4,6-trimethylthiophenol,2,4,6-trimethylbenzene-1-thiol,benzenethiol,2,4,6-trimethyl,2-mercaptomesitylene,2-sulphanylmesitylene,acmc-20aoem,2,4,6-trimethyl-benzene-1-thiol
284449
2,4,6-trimethylbenzenethiol
CC1=CC(=C(C(=C1)C)S)C
1541-10-2
C9H12S
152.255
MFCD00218475
PKANQZUPJCMBAK-UHFFFAOYSA-N
2,4,6-trimethylthiophenol,2,4,6-trimethylbenzene-1-thiol,benzenethiol,2,4,6-trimethyl,2-mercaptomesitylene,2-sulphanylmesitylene,acmc-20aoem,2,4,6-trimethyl-benzene-1-thiol
284449
2,4,6-trimethylbenzenethiol
CC1=CC(=C(C(=C1)C)S)C
Applications
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 25g
Formula Weight 152.25
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylthiophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning