Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triphenylpyrylium tetrafluoroborate ≥97%, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Plastic bottle
448-61-3
C23H17BF4O
396.192
MFCD00012001
VQYPWMWEJGDSTF-UHFFFAOYSA-N
2,4,6-triphenylpyrylium tetrafluoroborate,2,4,6-triphenyl-1??-pyran-1-ylium tetrafluoroborate,2,4,6-triphenylpyryliumtetrafluoroborate,pyrylium, 2,4,6-triphenyl-, tetrafluoroborate 1-,2,4,6-triphenyl-pyrylium tetrafluoroborate
9930615
2,4,6-triphenylpyrylium;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=C(C=C1)C2=CC(=[O+]C(=C2)C3=CC=CC=C3)C4=CC=CC=C4
448-61-3
C23H17BF4O
396.192
MFCD00012001
VQYPWMWEJGDSTF-UHFFFAOYSA-N
2,4,6-triphenylpyrylium tetrafluoroborate,2,4,6-triphenyl-1??-pyran-1-ylium tetrafluoroborate,2,4,6-triphenylpyryliumtetrafluoroborate,pyrylium, 2,4,6-triphenyl-, tetrafluoroborate 1-,2,4,6-triphenyl-pyrylium tetrafluoroborate
9930615
2,4,6-triphenylpyrylium;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC=C(C=C1)C2=CC(=[O+]C(=C2)C3=CC=CC=C3)C4=CC=CC=C4
Color Yellow
Infrared Spectrum Authentic
Quantity 25g
Packaging Plastic bottle
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 396.19
Physical Form Crystalline Powder
Percent Purity ≥97.0%
Chemical Name or Material 2, 4, 6-Triphenylpyrylium tetrafluoroborate
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning