Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-tris(tert-butylperoxy)-1,3,5-triazine, Acros Organics

Click to view available options
Quantity:
5g
854-44-4,108-88-3,103-23-1
C15H27N3O6
345.396
NSWXQQHHYHEEES-UHFFFAOYSA-N
2,4,6-tris tert-butylperoxy-1,3,5-triazine,tris tert-butylperoxy-1,3,5-triazine,1,3,5-triazine, 2,4,6-tris 1,1-dimethylethyl dioxy
2734923
2,4,6-tris(tert-butylperoxy)-1,3,5-triazine
CC(C)(C)OOC1=NC(=NC(=N1)OOC(C)(C)C)OOC(C)(C)C
854-44-4,108-88-3,103-23-1
C15H27N3O6
345.396
NSWXQQHHYHEEES-UHFFFAOYSA-N
2,4,6-tris tert-butylperoxy-1,3,5-triazine,tris tert-butylperoxy-1,3,5-triazine,1,3,5-triazine, 2,4,6-tris 1,1-dimethylethyl dioxy
2734923
2,4,6-tris(tert-butylperoxy)-1,3,5-triazine
CC(C)(C)OOC1=NC(=NC(=N1)OOC(C)(C)C)OOC(C)(C)C
Radical initiators
Quantity 5g
Color Colorless
Formula Weight 345.4
Physical Form Liquid
Percent Purity 33 to 37 wt%
Chemical Name or Material 2,4,6-Tris(tert-butylperoxy)-1,3,5-triazine

GHS H Statement:
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger