Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trivinylcyclotriboroxane-pyridine complex, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
95010-17-6
C11H14B3NO3
240.667
MFCD03839940
YLHJACXHRQQNQR-UHFFFAOYSA-N
2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1,trivinylboroxin pyridine complex,vinylboronic anhydride pyridine complex,2,4,6-trivinylcyclotriboroxane-pyridine,2,4,6-trivinylcyclotriboroxane pyridine,vinylboronic anhydride pyridine,2,4,6-trivinylcyclotriboroxane pyridine complex,2,4,6-trivinylcyclotriboroxane-pyridine complex,boroxin, triethenyl-, compd. with pyridine 1:1,pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
2734806
pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1
95010-17-6
C11H14B3NO3
240.667
MFCD03839940
YLHJACXHRQQNQR-UHFFFAOYSA-N
2,4,6-trivinyl-1,3,5,2,4,6-trioxatriborinane compound with pyridine 1:1,trivinylboroxin pyridine complex,vinylboronic anhydride pyridine complex,2,4,6-trivinylcyclotriboroxane-pyridine,2,4,6-trivinylcyclotriboroxane pyridine,vinylboronic anhydride pyridine,2,4,6-trivinylcyclotriboroxane pyridine complex,2,4,6-trivinylcyclotriboroxane-pyridine complex,boroxin, triethenyl-, compd. with pyridine 1:1,pyridine; triethenyl-1,3,5,2,4,6-trioxatriborinane
2734806
pyridine;2,4,6-tris(ethenyl)-1,3,5,2,4,6-trioxatriborinane
B1(OB(OB(O1)C=C)C=C)C=C.C1=CC=NC=C1
Quantity 250mg
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
Formula Weight 240.67
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 4, 6-Trivinylcyclotriboroxane-pyridine complex

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning