Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 96%

Click to view available options
Quantity:
1g
5g
25g
722-92-9
C9H7F6NO
259.151
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol,4-hexafluoro-2-hydroxyisopropyl aniline,2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol,2-4-aminophenyl hexafluoro-2-propanol,2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol,benzenemethanol,4-amino-a,a-bis trifluoromethyl,enamine_005408,4-hexafluoro-2-hydroxy-2-propyl aniline,4-hexafluoro-2-hydroxy-2-propyl-aniline,2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N
722-92-9
C9H7F6NO
259.151
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol,4-hexafluoro-2-hydroxyisopropyl aniline,2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol,2-4-aminophenyl hexafluoro-2-propanol,2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol,benzenemethanol,4-amino-a,a-bis trifluoromethyl,enamine_005408,4-hexafluoro-2-hydroxy-2-propyl aniline,4-hexafluoro-2-hydroxy-2-propyl-aniline,2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N
Melting Point 147°C to 152°C
Quantity 1g
Beilstein 1473737
Formula Weight 259.15
Percent Purity 96%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger