Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%, Thermo Scientific Chemicals

Catalog Number AC367810010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
722-92-9
C9H7F6NO
259.15
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-hexafluoro-2-hydroxyisopropyl aniline, 2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol, 2-4-aminophenyl hexafluoro-2-propanol, 2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol, benzenemethanol,4-amino-a,a-bis trifluoromethyl, enamine_005408, 4-hexafluoro-2-hydroxy-2-propyl aniline, 4-hexafluoro-2-hydroxy-2-propyl-aniline, 2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N
722-92-9
C9H7F6NO
259.15
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol, 4-hexafluoro-2-hydroxyisopropyl aniline, 2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol, 2-4-aminophenyl hexafluoro-2-propanol, 2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol, benzenemethanol,4-amino-a,a-bis trifluoromethyl, enamine_005408, 4-hexafluoro-2-hydroxy-2-propyl aniline, 4-hexafluoro-2-hydroxy-2-propyl-aniline, 2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 147°C to 152°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 259.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit