Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
722-92-9
C9H7F6NO
259.15
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol,4-hexafluoro-2-hydroxyisopropyl aniline,2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol,2-4-aminophenyl hexafluoro-2-propanol,2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol,benzenemethanol,4-amino-a,a-bis trifluoromethyl,enamine_005408,4-hexafluoro-2-hydroxy-2-propyl aniline,4-hexafluoro-2-hydroxy-2-propyl-aniline,2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N
722-92-9
C9H7F6NO
259.15
MFCD00039258
TZEJXCIGVMTMDY-UHFFFAOYSA-N
2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-ol,4-hexafluoro-2-hydroxyisopropyl aniline,2-4-aminophenyl-1,1,1,3,3,3-hexafluoro-2-propanol,2-4-aminophenyl hexafluoro-2-propanol,2-4-amino-phenyl-1,1,1,3,3,3-hexafluoro-propan-2-ol,benzenemethanol,4-amino-a,a-bis trifluoromethyl,enamine_005408,4-hexafluoro-2-hydroxy-2-propyl aniline,4-hexafluoro-2-hydroxy-2-propyl-aniline,2-4-aminophenyl hexafluoropropan-2-ol
577801
2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
C1=CC(=CC=C1C(C(F)(F)F)(C(F)(F)F)O)N
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 147°C to 152°C
Color Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 259.15
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.