Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole, 97%

Click to view available options
Quantity:
250mg
1g
749151-25-5
C16H14N2OS
282.361
FEJBCMDICNNALW-UHFFFAOYSA-N
2-4-aminophenyl-4-4-methoxyphenyl thiazole,4-4-4-methoxyphenyl-1,3-thiazol-2-yl aniline,4-4-4-methoxyphenyl thiazol-2-yl aniline
24261904
4-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]aniline
COC1=CC=C(C=C1)C2=CSC(=N2)C3=CC=C(C=C3)N
749151-25-5
C16H14N2OS
282.361
FEJBCMDICNNALW-UHFFFAOYSA-N
2-4-aminophenyl-4-4-methoxyphenyl thiazole,4-4-4-methoxyphenyl-1,3-thiazol-2-yl aniline,4-4-4-methoxyphenyl thiazol-2-yl aniline
24261904
4-[4-(4-methoxyphenyl)-1,3-thiazol-2-yl]aniline
COC1=CC=C(C=C1)C2=CSC(=N2)C3=CC=C(C=C3)N
Quantity 250mg
Color Brown
Formula Weight 282.36
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(4-Aminophenyl)-4-(4-methoxyphenyl)thiazole