Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-aminophenylthio)acetic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
104-18-7
C8H9NO2S
183.225
MFCD00025360
CTPIHHXCACYCIV-UHFFFAOYSA-N
4-aminophenylthio acetic acid,2-4-aminophenylthio acetic acid,4-aminophenyl sulfanyl acetic acid,acetic acid, 4-aminophenyl thio,p-aminophenylthioacetic acid,p-aminophenylmercaptoacetic acid,acetic acid,2-4-aminophenyl thio,p-aminothiophenoxyacetic acid,acetic acid, p-aminophenyl thio
66907
2-(4-aminophenyl)sulfanylacetic acid
C1=CC(=CC=C1N)SCC(=O)O
104-18-7
C8H9NO2S
183.225
MFCD00025360
CTPIHHXCACYCIV-UHFFFAOYSA-N
4-aminophenylthio acetic acid,2-4-aminophenylthio acetic acid,4-aminophenyl sulfanyl acetic acid,acetic acid, 4-aminophenyl thio,p-aminophenylthioacetic acid,p-aminophenylmercaptoacetic acid,acetic acid,2-4-aminophenyl thio,p-aminothiophenoxyacetic acid,acetic acid, p-aminophenyl thio
66907
2-(4-aminophenyl)sulfanylacetic acid
C1=CC(=CC=C1N)SCC(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Khaki
Quantity 1g
Melting Point 193°C to 203°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula NH2C6H4SCH2COOH
Formula Weight 183.23
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-(4-Aminophenylthio)acetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning