Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Biphenylyl)ethylamine, 98%

Manufacturer:  Alfa Aesar™ H2765403

Quantity 1g

 View more versions of this product

Catalog No. AAH2765403

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is used in OLED materials and in pharmaceutical intermediate. It is a potential cardiovascular and central nervous system agent.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool dry conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

17027-51-9
197.28
2-(4-Biphenylyl)ethylamine
C14H15N
MFCD01310786

Specifications

17027-51-9
197.28
50° to 54°C
>110°C (230°F)
98%
C14H15N
MFCD01310786
1g
2-(4-Biphenylyl)ethylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit