Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4'-Bipyridine, 97%

Manufacturer:  Alfa Aesar™ H27366MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27366MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

581-47-5
156.19
UN2811
2,4'-Bipyridine
C10H8N2
MFCD00006217
2,4'-Bipyridyl; 2,4'-Dipyridyl

Specifications

581-47-5
156.19
UN2811
281°C
250mg
97%
C10H8N2
MFCD00006217
59° to 63°C
2,4'-Bipyridyl; 2,4'-Dipyridyl
2,4'-Bipyridine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit