Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Bis(trifluoromethyl)benzamide, 97+%

Click to view available options
Quantity:
1g
5g
53130-45-3
C9H5F6NO
257.135
MFCD00083536
UFXIIQGMMJQZPG-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzamide,acmc-1an7n,2,4-bis trifluoromethyl benzamid,4,6-bis-trifluoromethyl-benzamide,benzamide,2,4-bis trifluoromethyl
2736099
2,4-bis(trifluoromethyl)benzamide
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)N
53130-45-3
C9H5F6NO
257.135
MFCD00083536
UFXIIQGMMJQZPG-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzamide,acmc-1an7n,2,4-bis trifluoromethyl benzamid,4,6-bis-trifluoromethyl-benzamide,benzamide,2,4-bis trifluoromethyl
2736099
2,4-bis(trifluoromethyl)benzamide
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)N
Quantity 5g
Formula Weight 257.14
Percent Purity ≥97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning