Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Bis(trifluoromethyl)benzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
140690-56-8
C9H5BrF6
307.033
MFCD00010306
SWFFFUJOWAJJCH-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzyl bromide,1-bromomethyl-2,4-bis trifluoromethyl benzene,2,4-bis trifluoromethyl benzylbromide,benzene, 1-bromomethyl-2,4-bis trifluoromethyl,2,4-bis trifluoromethyl-1-bromomethyl benzene,pubchem4879,acmc-1c0tm,2,4-bis-trifluoromethyl-benzylbromide,2,4-bis-trifluoromethyl benzyl bromide,2,4-bis-trifluoromethyl-benzyl bromide
518871
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr
140690-56-8
C9H5BrF6
307.033
MFCD00010306
SWFFFUJOWAJJCH-UHFFFAOYSA-N
2,4-bis trifluoromethyl benzyl bromide,1-bromomethyl-2,4-bis trifluoromethyl benzene,2,4-bis trifluoromethyl benzylbromide,benzene, 1-bromomethyl-2,4-bis trifluoromethyl,2,4-bis trifluoromethyl-1-bromomethyl benzene,pubchem4879,acmc-1c0tm,2,4-bis-trifluoromethyl-benzylbromide,2,4-bis-trifluoromethyl benzyl bromide,2,4-bis-trifluoromethyl-benzyl bromide
518871
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr
Boiling Point 112°C (0.2mmHg)
Quantity 5g
Flash Point 92°C (197°F)
UN Number UN3265
Beilstein 7995492
Formula Weight 307.04
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzyl bromide

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.