Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)-1-phenylbenzimidazole, 97%

Click to view available options
Quantity:
250mg
1g
5g
2620-76-0
C19H13BrN2
349.231
MFCD09261342
DXRLALXPCIOIDK-UHFFFAOYSA-N
2-4-bromophenyl-1-phenyl-1h-benzoimidazole,2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole,2-4-bromophenyl-1-phenyl-1h-benzimidazole,2-4-bromophenyl-1-phenylbenzimidazole,2-4-bromophenyl-1-phenyl-1h-benzo d imidazole,1h-benzimidazole, 2-4-bromophenyl-1-phenyl,1-phenyl-2-4-bromophenyl benzimidazole,2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole,2-4-bromophenyl-1-phenyl-1,3-benzodiazole,zlchem 100
23094070
2-(4-bromophenyl)-1-phenylbenzimidazole
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br
2620-76-0
C19H13BrN2
349.231
MFCD09261342
DXRLALXPCIOIDK-UHFFFAOYSA-N
2-4-bromophenyl-1-phenyl-1h-benzoimidazole,2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole,2-4-bromophenyl-1-phenyl-1h-benzimidazole,2-4-bromophenyl-1-phenylbenzimidazole,2-4-bromophenyl-1-phenyl-1h-benzo d imidazole,1h-benzimidazole, 2-4-bromophenyl-1-phenyl,1-phenyl-2-4-bromophenyl benzimidazole,2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole,2-4-bromophenyl-1-phenyl-1,3-benzodiazole,zlchem 100
23094070
2-(4-bromophenyl)-1-phenylbenzimidazole
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br
Quantity 1g
Color White
Melting Point 162°C to 166°C
Beilstein 617138
Formula Weight 349.23
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenyl)-1-phenylbenzimidazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning