Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)-2-propanol, 98%

Click to view available options
Quantity:
250mg
1g
2077-19-2
C9H11BrO
215.09
MFCD11870096
AOGYBHJTXLXRSM-UHFFFAOYSA-N
2-4-bromophenyl propan-2-ol,2-4-bromophenyl-2-propanol,acmc-209fdx,2-4-bromophenyl propan-2-o1,2-4-bromophenyl-propan-2-ol,2-4-bromo-phenyl propan-2-ol,2-4-bromo-phenyl-propan-2-ol,p-bromo-alpha,alpha-dimethylbenzyl alcohol,4-bromo-alpha,alpha-dimethylbenzyl alcohol
12681920
2-(4-bromophenyl)propan-2-ol
CC(C)(C1=CC=C(C=C1)Br)O
2077-19-2
C9H11BrO
215.09
MFCD11870096
AOGYBHJTXLXRSM-UHFFFAOYSA-N
2-4-bromophenyl propan-2-ol,2-4-bromophenyl-2-propanol,acmc-209fdx,2-4-bromophenyl propan-2-o1,2-4-bromophenyl-propan-2-ol,2-4-bromo-phenyl propan-2-ol,2-4-bromo-phenyl-propan-2-ol,p-bromo-alpha,alpha-dimethylbenzyl alcohol,4-bromo-alpha,alpha-dimethylbenzyl alcohol
12681920
2-(4-bromophenyl)propan-2-ol
CC(C)(C1=CC=C(C=C1)Br)O
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.

Solubility
Slightly soluble in water (1.4 g/L) (25°C)

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Refractive Index 1.5520
Density 1.356g/mL
Color White
Melting Point 40°C to 45°C
Quantity 1g
Boiling Point 75°C to 85°C (0.1mmHg)
Beilstein 1935066
Formula Weight 215.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-(4-Bromophenyl)-2-propanol
Show More Show Less