Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)acetamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
5g
422560-40-5
C8H9BrN2O
229.077
MFCD05662829
WHICEIACQGPSMU-UHFFFAOYSA-N
2-4-bromophenyl-1-hydroxyimino ethylamine,2-4-bromo-phenyl-n-hydroxy-acetamidine,e-2-4-bromophenyl-n'-hydroxyethanimidamide,z-2-4-bromophenyl-n'-hydroxyethanimidamide
6029814
2-(4-bromophenyl)-N'-hydroxyethanimidamide
C1=CC(=CC=C1CC(=NO)N)Br
422560-40-5
C8H9BrN2O
229.077
MFCD05662829
WHICEIACQGPSMU-UHFFFAOYSA-N
2-4-bromophenyl-1-hydroxyimino ethylamine,2-4-bromo-phenyl-n-hydroxy-acetamidine,e-2-4-bromophenyl-n'-hydroxyethanimidamide,z-2-4-bromophenyl-n'-hydroxyethanimidamide
6029814
2-(4-bromophenyl)-N'-hydroxyethanimidamide
C1=CC(=CC=C1CC(=NO)N)Br
Quantity 5g
Color White
Melting Point 132°C to 134°C
Formula Weight 229.07
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenyl)acetamidoxime