Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide, 96%

Click to view available options
Quantity:
5g
25g
32022-38-1
C9H11ClN2O2
214.649
MFCD00082720
JXQVZCTYFYWQGN-UHFFFAOYSA-N
2-4-chloro-2-methylphenoxy acetohydrazide,2-4-chloro-2-methylphenoxy acetic acid hydrazide,acetic acid, 4-chloro-o-tolyl oxy-, hydrazide,acmc-20aopt,4-chloro-o-tolyl oxy acetic acid hydrazide,2-4-chloro-2-methyl-phenoxy acetohydrazide,2-4-chloro-3-methylphenoxy acetic hydrazide,2-4-chloro-o-tolyloxy acetic acid hydrazide,2-methyl-4-chlorophenoxyacetic acid hydrazide,4-chloro-2-methylphenoxy acetic acid hydrazid
149463
2-(4-chloro-2-methylphenoxy)acetohydrazide
CC1=C(C=CC(=C1)Cl)OCC(=O)NN
32022-38-1
C9H11ClN2O2
214.649
MFCD00082720
JXQVZCTYFYWQGN-UHFFFAOYSA-N
2-4-chloro-2-methylphenoxy acetohydrazide,2-4-chloro-2-methylphenoxy acetic acid hydrazide,acetic acid, 4-chloro-o-tolyl oxy-, hydrazide,acmc-20aopt,4-chloro-o-tolyl oxy acetic acid hydrazide,2-4-chloro-2-methyl-phenoxy acetohydrazide,2-4-chloro-3-methylphenoxy acetic hydrazide,2-4-chloro-o-tolyloxy acetic acid hydrazide,2-methyl-4-chlorophenoxyacetic acid hydrazide,4-chloro-2-methylphenoxy acetic acid hydrazid
149463
2-(4-chloro-2-methylphenoxy)acetohydrazide
CC1=C(C=CC(=C1)Cl)OCC(=O)NN
Melting Point 149°C to 151°C
Quantity 25g
Beilstein 983137
Formula Weight 214.65
Percent Purity 96%
Chemical Name or Material 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning