Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
882-09-7
C10H11ClO3
214.65
MFCD00004192
TXCGAZHTZHNUAI-UHFFFAOYSA-N
clofibric acid,2-4-chlorophenoxy-2-methylpropanoic acid,chlorofibrinic acid,clofibrinic acid,chlorfibrinic acid,clofibrin,chlorophibrinic acid,pcib,2-4-chlorophenoxy-2-methylpropionic acid,clofibrate free acid
2797
CHEBI:34648
2-(4-chlorophenoxy)-2-methylpropanoic acid
CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl
882-09-7
C10H11ClO3
214.65
MFCD00004192
TXCGAZHTZHNUAI-UHFFFAOYSA-N
clofibric acid,2-4-chlorophenoxy-2-methylpropanoic acid,chlorofibrinic acid,clofibrinic acid,chlorfibrinic acid,clofibrin,chlorophibrinic acid,pcib,2-4-chlorophenoxy-2-methylpropionic acid,clofibrate free acid
2797
CHEBI:34648
2-(4-chlorophenoxy)-2-methylpropanoic acid
CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl
Infrared Spectrum Authentic
Color Brown to Yellow
Melting Point 120.0°C to 123.0°C
Packaging Plastic bottle
Loss on Drying 1% max.
Quantity 250g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H4OC(CH3)2CO2H
Merck Index 15, 2371
Formula Weight 214.65
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 212-925-9

RTECSNumber : UE9455000

TSCA : TSCA