Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Chlorophenyl)-1,3-indanedione, 97%

Click to view available options
Quantity:
1g
1146-99-2
C15H9ClO2
256.685
MFCD00239203
NJDUWAXIURWWLN-UHFFFAOYSA-N
clorindione,chlorindione,chlophenadione,chlorindionum,indaliton,chlor-athrombon,chlorphenylindandione,2-4-chlorophenyl indane-1,3-dione,2-4-chlorophenyl-1h-indene-1,3 2h-dione,clorindione inn:ban
70846
2-(4-chlorophenyl)indene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)C3=CC=C(C=C3)Cl
1146-99-2
C15H9ClO2
256.685
MFCD00239203
NJDUWAXIURWWLN-UHFFFAOYSA-N
clorindione,chlorindione,chlophenadione,chlorindionum,indaliton,chlor-athrombon,chlorphenylindandione,2-4-chlorophenyl indane-1,3-dione,2-4-chlorophenyl-1h-indene-1,3 2h-dione,clorindione inn:ban
70846
2-(4-chlorophenyl)indene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)C3=CC=C(C=C3)Cl
Quantity 1g
UN Number UN2811
Formula Weight 256.69
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenyl)-1,3-indanedione

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-553-2

TSCA : No

Recommended Storage : Ambient temperatures