Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenyl)-2-methylpropionic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
6258-30-6
C10H11ClO2
198.646
MFCD00044814
SSFDAZXGUKDEAH-UHFFFAOYSA-N
2-4-chlorophenyl-2-methylpropanoic acid,2-4-chlorophenyl-2-methylpropionic acid,4-chloro-alpha,alpha-dimethylphenylacetic acid,2-p-chlorophenyl-2-methylpropionic acid,4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid,acmc-1bhp2,p-chlorophenylisobutyric acid,2-methyl-2-4-chlorophenyl propanoic acid,4-chloro-a,a-dimethylphenylacetic acid,2-4-chloro phenyl-2-methylpropanoic acid
80406
2-(4-chlorophenyl)-2-methylpropanoic acid
CC(C)(C1=CC=C(C=C1)Cl)C(=O)O
6258-30-6
C10H11ClO2
198.646
MFCD00044814
SSFDAZXGUKDEAH-UHFFFAOYSA-N
2-4-chlorophenyl-2-methylpropanoic acid,2-4-chlorophenyl-2-methylpropionic acid,4-chloro-alpha,alpha-dimethylphenylacetic acid,2-p-chlorophenyl-2-methylpropionic acid,4-chloro-.alpha.,.alpha.-dimethylphenylacetic acid,acmc-1bhp2,p-chlorophenylisobutyric acid,2-methyl-2-4-chlorophenyl propanoic acid,4-chloro-a,a-dimethylphenylacetic acid,2-4-chloro phenyl-2-methylpropanoic acid
80406
2-(4-chlorophenyl)-2-methylpropanoic acid
CC(C)(C1=CC=C(C=C1)Cl)C(=O)O
Quantity 10g
Color White
Packaging Glass bottle
Melting Point 122°C to 127°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 198.65
Physical Form Fine Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-(4-Chlorophenyl)-2-methylpropionic acid, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.