Alfa Aesar™ 2-(4-Chlorophenylthio)-6-fluorobenzonitrile, 97%

Manufacturer: Alfa Aesar™ B2523014

Quantity 25g

 View more versions of this product

Catalog No. AAB2523014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 175204-12-3
    Molecular Formula C13H7ClFNS
    Formula Weight 263.72
    MDL Number MFCD00068208
    Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile

    Specifications

    Formula Weight 263.72
    Molecular Formula C13H7ClFNS
    MDL Number MFCD00068208
    CAS 175204-12-3
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 68° to 71°C
    Quantity 25g
    Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile
    Assay 97%

    GHS H Statement
    H301-H311-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit