Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Chlorophenylthio)-6-fluorobenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
175204-12-3
C13H7ClFNS
263.714
MFCD00068208
KFJFXRCXNMCNBN-UHFFFAOYSA-N
2-fluoro-6-4-chlorophenylthio benzonitrile,2-4-chlorophenyl thio-6-fluorobenzonitrile,2-4-chlorophenyl sulfanyl-6-fluorobenzonitrile,2-4-chlorophenylthio-6-fluorobenzonitrile,2-4-chlorophenylthio-6-fluorobenzenecarbonitrile,acmc-20anur,maybridge3_004320,2-4-chlorophenylsulfanyl-6-fluorobenzonitrile
2737296
2-(4-chlorophenyl)sulfanyl-6-fluorobenzonitrile
C1=CC(=C(C(=C1)SC2=CC=C(C=C2)Cl)C#N)F
175204-12-3
C13H7ClFNS
263.714
MFCD00068208
KFJFXRCXNMCNBN-UHFFFAOYSA-N
2-fluoro-6-4-chlorophenylthio benzonitrile,2-4-chlorophenyl thio-6-fluorobenzonitrile,2-4-chlorophenyl sulfanyl-6-fluorobenzonitrile,2-4-chlorophenylthio-6-fluorobenzonitrile,2-4-chlorophenylthio-6-fluorobenzenecarbonitrile,acmc-20anur,maybridge3_004320,2-4-chlorophenylsulfanyl-6-fluorobenzonitrile
2737296
2-(4-chlorophenyl)sulfanyl-6-fluorobenzonitrile
C1=CC(=C(C(=C1)SC2=CC=C(C=C2)Cl)C#N)F
Melting Point 68°C to 71°C
Quantity 1g
Formula Weight 263.72
Percent Purity 97%
Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger