Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
126519-89-9,75-09-2
C11H17ClO5SSi
324.85
MFCD00054774
NYIDSUMRGUILGR-UHFFFAOYSA-N
2-4-chlorosulfonylphenyl ethyltrimethoxysilane,4-2-trimethoxysilylethyl benzenesulfonyl chloride,4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride,benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl,2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid,4-2-trimethoxysilyl ethyl benzenesulfonyl chloride,acmc-20ete8,4-chlorosulfonyl phenethyltrimethoxysilane
2733807
4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
CO[Si](CCC1=CC=C(C=C1)S(=O)(=O)Cl)(OC)OC
126519-89-9,75-09-2
C11H17ClO5SSi
324.85
MFCD00054774
NYIDSUMRGUILGR-UHFFFAOYSA-N
2-4-chlorosulfonylphenyl ethyltrimethoxysilane,4-2-trimethoxysilylethyl benzenesulfonyl chloride,4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride,benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl,2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid,4-2-trimethoxysilyl ethyl benzenesulfonyl chloride,acmc-20ete8,4-chlorosulfonyl phenethyltrimethoxysilane
2733807
4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
CO[Si](CCC1=CC=C(C=C1)S(=O)(=O)Cl)(OC)OC
Infrared Spectrum Authentic
Quantity 5g
Color Yellow
Boiling Point >40.0°C
Packaging Glass bottle
CAS Min % 45.0,45.0
CAS Max % 55.0,55.0
Solubility Information Solubility in water: reacts
Formula Weight 324.85
Physical Form Solution
Percent Purity 45 to 55% (solids)
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less

GHS H Statement
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 279-267-2

TSCA : TSCA