Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
126519-89-9,75-09-2
C11H17ClO5SSi
324.847
MFCD00054774
NYIDSUMRGUILGR-UHFFFAOYSA-N
2-4-chlorosulfonylphenyl ethyltrimethoxysilane,4-2-trimethoxysilylethyl benzenesulfonyl chloride,4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride,benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl,2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid,4-2-trimethoxysilyl ethyl benzenesulfonyl chloride,acmc-20ete8,4-chlorosulfonyl phenethyltrimethoxysilane
2733807
4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
CO[Si](CCC1=CC=C(C=C1)S(=O)(=O)Cl)(OC)OC
126519-89-9,75-09-2
C11H17ClO5SSi
324.847
MFCD00054774
NYIDSUMRGUILGR-UHFFFAOYSA-N
2-4-chlorosulfonylphenyl ethyltrimethoxysilane,4-2-trimethoxysilylethyl benzenesulfonyl chloride,4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride,benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl,2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid,4-2-trimethoxysilyl ethyl benzenesulfonyl chloride,acmc-20ete8,4-chlorosulfonyl phenethyltrimethoxysilane
2733807
4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
CO[Si](CCC1=CC=C(C=C1)S(=O)(=O)Cl)(OC)OC
Infrared Spectrum Authentic
Quantity 25g
Boiling Point >40.0°C
Color Yellow
Packaging Glass bottle
CAS Min % 45.0,45.0
CAS Max % 55.0,55.0
Formula Weight 324.85
Physical Form Solution
Percent Purity 45 to 55% (solids)
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 279-267-2

TSCA : TSCA