Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,4-Diamino-6-hydroxypyrimidine, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
GTP cyclohydrolase I (GCH1) inhibitor. Prevents the de novo synthesis of tetrahydrobiopterin (BH4) and thus suppresses the activity of NO synthase. Exhibits more potent inhibition of GCH1 in the presence of GFRP (GTP cyclohydrolase feedback-regulatory protein).
Specifications
Quantity | 50 mg |
Formula Weight | 126.12 |
Percent Purity | >99% |
Chemical Name or Material | 2, 4-Diamino-6-hydroxypyrimidine |
Safety and Handling
Recommended Storage : Desiccate at +4°C