Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Diamino-6-methyl-1,3,5-triazine, 96%

Click to view available options
Quantity:
100g
500g
542-02-9
C4H7N5
125.135
MFCD00023192
NJYZCEFQAIUHSD-UHFFFAOYSA-N
acetoguanamine,2,4-diamino-6-methyl-1,3,5-triazine,1,3,5-triazine-2,4-diamine, 6-methyl,2-methyl-4,6-diamino-1,3,5-triazine,2,4-diamino-6-methyl-s-triazine,6-methyl-1,3,5-triazine-2,4-diyldiamine,s-triazine, 2,4-diamino-6-methyl,acetoguanamin,ccris 8909,dsstox_cid_9186
10949
CHEBI:72475
6-methyl-1,3,5-triazine-2,4-diamine
CC1=NC(=NC(=N1)N)N
542-02-9
C4H7N5
125.135
MFCD00023192
NJYZCEFQAIUHSD-UHFFFAOYSA-N
acetoguanamine,2,4-diamino-6-methyl-1,3,5-triazine,1,3,5-triazine-2,4-diamine, 6-methyl,2-methyl-4,6-diamino-1,3,5-triazine,2,4-diamino-6-methyl-s-triazine,6-methyl-1,3,5-triazine-2,4-diyldiamine,s-triazine, 2,4-diamino-6-methyl,acetoguanamin,ccris 8909,dsstox_cid_9186
10949
CHEBI:72475
6-methyl-1,3,5-triazine-2,4-diamine
CC1=NC(=NC(=N1)N)N
Melting Point ~280°C (decomposition)
Quantity 100g
Beilstein 118348
Formula Weight 125.13
Percent Purity 96%
Chemical Name or Material 2,4-Diamino-6-methyl-1,3,5-triazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA