Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminophenol dihydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
137-09-7
C6H8N22HCl
197.07
MFCD00012979
KQEIJFWAXDQUPR-UHFFFAOYSA-N
2,4-diaminophenol dihydrochloride,amidol,dianol,acrol,2,4-diaminophenol hcl,2,4-diaminophenol.2hcl,phenol, 2,4-diamino-, dihydrochloride,2,4-diaminophenol hydrochloride,diamidophenol hydrochloride,unii-t1iyy101nx
8715
2,4-diaminophenol;dihydrochloride
C1=CC(=C(C=C1N)N)O.Cl.Cl
137-09-7
C6H8N22HCl
197.07
MFCD00012979
KQEIJFWAXDQUPR-UHFFFAOYSA-N
2,4-diaminophenol dihydrochloride,amidol,dianol,acrol,2,4-diaminophenol hcl,2,4-diaminophenol.2hcl,phenol, 2,4-diamino-, dihydrochloride,2,4-diaminophenol hydrochloride,diamidophenol hydrochloride,unii-t1iyy101nx
8715
2,4-diaminophenol;dihydrochloride
C1=CC(=C(C=C1N)N)O.Cl.Cl
Packaging Glass bottle
Melting Point 222°C
Color Beige to Gray-Green
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula (H2N)2C6H3OH·2HCl
Beilstein 13, 550
Merck Index 14, 2983
Solubility Information Solubility in water: 27.5g/100mL (15°C). Other solubilities: slightly soluble in alcohol and ether
Formula Weight 197.07
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 4-Diaminophenol dihydrochloride
Show More Show Less

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-279-4