Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Diaminothiazole hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
72792-54-2
C3H6ClN3S
151.612
MFCD01438626
PQAUAFCGXNRKIU-UHFFFAOYSA-N
thiazole-2,4-diamine hydrochloride,1,3-thiazole-2,4-diamine hydrochloride,thiazole-2,4-diamine hcl,2-thiazolamine, 4,5-dihydro-4-imino-, monohydrochloride,4-thiazolamine, 2,5-dihydro-2-imino-, hydrochloride 1:1,1,3-thiazole-2,4-diamine hydrochloride 1:1,2,4-diaminothiazole, monohydrochloride,1,3-thiazole-2,4-diaminehydrochloride,1,3-thiazole-2,4-diamine hydrochloride 1:1
118994
1,3-thiazole-2,4-diamine;hydrochloride
C1=C(N=C(S1)N)N.Cl
72792-54-2
C3H6ClN3S
151.612
MFCD01438626
PQAUAFCGXNRKIU-UHFFFAOYSA-N
thiazole-2,4-diamine hydrochloride,1,3-thiazole-2,4-diamine hydrochloride,thiazole-2,4-diamine hcl,2-thiazolamine, 4,5-dihydro-4-imino-, monohydrochloride,4-thiazolamine, 2,5-dihydro-2-imino-, hydrochloride 1:1,1,3-thiazole-2,4-diamine hydrochloride 1:1,2,4-diaminothiazole, monohydrochloride,1,3-thiazole-2,4-diaminehydrochloride,1,3-thiazole-2,4-diamine hydrochloride 1:1
118994
1,3-thiazole-2,4-diamine;hydrochloride
C1=C(N=C(S1)N)N.Cl
Quantity 1g
Beilstein 3691448
Formula Weight 151.61
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2,4-Diaminothiazole hydrochloride