Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Diaminotoluene, 98%

Click to view available options
Quantity:
250g
1000g
95-80-7
C7H10N2
122.171
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene,2,4-toluenediamine,developer t,toluene-2,4-diamine,1,3-benzenediamine, 4-methyl,developer mtd,m-toluenediamine,fourrine m,m-tolylenediamine,benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=C(C=C(C=C1)N)N
95-80-7
C7H10N2
122.171
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene,2,4-toluenediamine,developer t,toluene-2,4-diamine,1,3-benzenediamine, 4-methyl,developer mtd,m-toluenediamine,fourrine m,m-tolylenediamine,benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=C(C=C(C=C1)N)N
Quantity 1000g
Flash Point 169°C (336°F)
Melting Point 96°C to 101°C
Odor Recognizable
Boiling Point 283°C to 285°C
UN Number UN1709
Beilstein 2205839
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,4-Diaminotoluene

GHS H Statement
H301-H350-H341-H361-H373-H312-H317
Toxic if swallowed.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P309-P310
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2,4-TOLUYLENEDIAMINE, SOLID

RTECSNumber : XS9625000

TSCA : TSCA