Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminotoluene 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
Packaging:
Glass bottle
95-80-7
C7H10N2
122.17
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene,2,4-toluenediamine,developer t,toluene-2,4-diamine,1,3-benzenediamine, 4-methyl,developer mtd,m-toluenediamine,fourrine m,m-tolylenediamine,benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=C(C=C(C=C1)N)N
95-80-7
C7H10N2
122.17
MFCD00007804
VOZKAJLKRJDJLL-UHFFFAOYSA-N
2,4-diaminotoluene,2,4-toluenediamine,developer t,toluene-2,4-diamine,1,3-benzenediamine, 4-methyl,developer mtd,m-toluenediamine,fourrine m,m-tolylenediamine,benzofur mt
7261
CHEBI:34237
4-methylbenzene-1,3-diamine
CC1=C(C=C(C=C1)N)N
Melting Point 97.0°C to 101.0°C
Infrared Spectrum Authentic
Boiling Point 283.0°C to 285.0°C
Quantity 1kg
Flash Point 149°C
Packaging Glass bottle
Color Brown to Gray
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H3(NH2)2
Beilstein 13, 124
Solubility Information Solubility in water: 50g/L (25 c). Other solubilities: soluble in alcohol, ether, chloroform and, benzene
Formula Weight 122.17
Physical Form Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 2, 4-Diaminotoluene, 98%
Show More Show Less

GHS H Statement
May cause cancer. Suspected of causing genetic defects. Suspected of damaging fertility. Toxic if swallowed. Harmful in contact with skin. May cause damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

Danger

EINECSNumber : 202-453-1

RTECSNumber : XS9625000

TSCA : TSCA