Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromo-1-fluorobenzene, 98%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
1435-53-6
C6H3BrF
253.9
MFCD00000283
MKHDDTWHDFVYDQ-UHFFFAOYSA-N
2,4-dibromofluorobenzene,1,3-dibromo-4-fluorobenzene,1-fluoro-2,4-dibromobenzene,benzene, 2,4-dibromo-1-fluoro,pubchem3446,acmc-209cqz,1,3-dibromo-5-fluoro,intermediates-zcf02097,ksc494g9j
15034
2,4-dibromo-1-fluorobenzene
C1=CC(=C(C=C1Br)Br)F
1435-53-6
C6H3BrF
253.9
MFCD00000283
MKHDDTWHDFVYDQ-UHFFFAOYSA-N
2,4-dibromofluorobenzene,1,3-dibromo-4-fluorobenzene,1-fluoro-2,4-dibromobenzene,benzene, 2,4-dibromo-1-fluoro,pubchem3446,acmc-209cqz,1,3-dibromo-5-fluoro,intermediates-zcf02097,ksc494g9j
15034
2,4-dibromo-1-fluorobenzene
C1=CC(=C(C=C1Br)Br)F
Boiling Point 105°C to 110°C (22.0mmHg)
Flash Point 92°C
Specific Gravity 2.047
Refractive Index 1.5820 to 1.584
Quantity 25mL
Infrared Spectrum Authentic
Color Colorless
Packaging Glass bottle
Density 2.0470g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Br2C6H3F
Solubility Information Solubility in water: insoluble
Formula Weight 253.9
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dibromo-1-fluorobenzene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.