Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,4-Dibromo-6-fluorophenol, 97%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Odor | Phenol-like |
Melting Point | 30°C to 33°C |
Quantity | 1g |
Formula Weight | 269.91 |
Percent Purity | 97% |
Chemical Name or Material | 2,4-Dibromo-6-fluorophenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures