Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromo-6-nitroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
827-23-6
C6H4Br2N2O2
295.918
MFCD00012107
OBTUHOVIVLDLLD-UHFFFAOYSA-N
benzenamine, 2,4-dibromo-6-nitro,2,4-dibromo-6-nitrophenylamine,2,4-dibromo-6-nitrobenzenamine,acmc-209pq0,2-nitro-4,6-dibromoaniline,4,6-dibromo-2-nitroaniline,2,4-dibromo-6-nitro-aniline,2,4-dibromo-6-nitro-phenylamine,aniline, 2,4-dibromo-6-nitro,benzenamine,2,4-dibromo-6-nitro
70009
2,4-dibromo-6-nitroaniline
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br
827-23-6
C6H4Br2N2O2
295.918
MFCD00012107
OBTUHOVIVLDLLD-UHFFFAOYSA-N
benzenamine, 2,4-dibromo-6-nitro,2,4-dibromo-6-nitrophenylamine,2,4-dibromo-6-nitrobenzenamine,acmc-209pq0,2-nitro-4,6-dibromoaniline,4,6-dibromo-2-nitroaniline,2,4-dibromo-6-nitro-aniline,2,4-dibromo-6-nitro-phenylamine,aniline, 2,4-dibromo-6-nitro,benzenamine,2,4-dibromo-6-nitro
70009
2,4-dibromo-6-nitroaniline
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br
Packaging Glass bottle
Quantity 5g
Color Orange-Yellow
Melting Point 129°C to 133°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 295.92
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Dibromo-6-nitroaniline, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.