Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dibromoanisole, 98+%

Click to view available options
Quantity:
25g
100g
21702-84-1
C7H6Br2O
265.932
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
2,4-dibromoanisole,benzene, 2,4-dibromo-1-methoxy,benzene,2,4-dibromo-1-methoxy,pubchem3847,acmc-209fnf,maybridge1_003667,chemwish ic06728,2,4-dibromo-1-methoxy-benzene,xgxugxpkrbqins-uhfffaoysa,#
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br
21702-84-1
C7H6Br2O
265.932
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
2,4-dibromoanisole,benzene, 2,4-dibromo-1-methoxy,benzene,2,4-dibromo-1-methoxy,pubchem3847,acmc-209fnf,maybridge1_003667,chemwish ic06728,2,4-dibromo-1-methoxy-benzene,xgxugxpkrbqins-uhfffaoysa,#
27011
2,4-dibromo-1-methoxybenzene
COC1=C(C=C(C=C1)Br)Br
Boiling Point 82°C to 88°C (0.3mmHg)
Melting Point 60°C to 64°C
Quantity 25g
Beilstein 2045290
Formula Weight 265.93
Percent Purity ≥98%
Chemical Name or Material 2,4-Dibromoanisole

EINECSNumber : 244-536-5

TSCA : No

Recommended Storage : Ambient temperatures