Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dibromophenol, 98%

Click to view available options
Quantity:
25g
100g
500g
615-58-7
C6H4Br2O
251.905
MFCD00002149
FAXWFCTVSHEODL-UHFFFAOYSA-N
phenol, 2,4-dibromo,2,4-dibromo-phenol,2,4-dibromphenol,unii-ia75t5c9tg,2,4-bis bromanyl phenol,ia75t5c9tg,phenol,4-dibromo,emery 9331,acmc-1bga9,2,4-dibromophenol
12005
CHEBI:34238
2,4-dibromophenol
C1=CC(=C(C=C1Br)Br)O
615-58-7
C6H4Br2O
251.905
MFCD00002149
FAXWFCTVSHEODL-UHFFFAOYSA-N
phenol, 2,4-dibromo,2,4-dibromo-phenol,2,4-dibromphenol,unii-ia75t5c9tg,2,4-bis bromanyl phenol,ia75t5c9tg,phenol,4-dibromo,emery 9331,acmc-1bga9,2,4-dibromophenol
12005
CHEBI:34238
2,4-dibromophenol
C1=CC(=C(C=C1Br)Br)O
Melting Point 35°C to 38°C
Flash Point >110°C (230°F)
Quantity 25g
Odor Phenol-like
Boiling Point 239°C
UN Number UN2811
Beilstein 1861291
Formula Weight 251.92
Percent Purity 98%
Chemical Name or Material 2,4-Dibromophenol

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H300-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 210-436-5

RTECSNumber : SK8010000

TSCA : Yes

Recommended Storage : Ambient temperatures