Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dibromopropiophenone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
38786-67-3
C9H8Br2O
291.97
MFCD00209571
GKALOSTUZMFUQB-UHFFFAOYSA-N
2,4'-dibromopropiophenone,2-bromo-1-4-bromophenyl propan-1-one,1-propanone, 2-bromo-1-4-bromophenyl,2,4-dibromopropiophenone,2,4'-dibromopropiophenon,acmc-1ago1,alpha,4'-dibromopropiophenone,1-propanone,2-bromo-1-4-bromophenyl,2-bromo-1-4-bromophenyl-1-propanone #
603907
2-bromo-1-(4-bromophenyl)propan-1-one
CC(C(=O)C1=CC=C(C=C1)Br)Br
38786-67-3
C9H8Br2O
291.97
MFCD00209571
GKALOSTUZMFUQB-UHFFFAOYSA-N
2,4'-dibromopropiophenone,2-bromo-1-4-bromophenyl propan-1-one,1-propanone, 2-bromo-1-4-bromophenyl,2,4-dibromopropiophenone,2,4'-dibromopropiophenon,acmc-1ago1,alpha,4'-dibromopropiophenone,1-propanone,2-bromo-1-4-bromophenyl,2-bromo-1-4-bromophenyl-1-propanone #
603907
2-bromo-1-(4-bromophenyl)propan-1-one
CC(C(=O)C1=CC=C(C=C1)Br)Br
Color Orange to White
Melting Point 84°C to 88°C
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H4COCH(Br)CH3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylacetate,ethanol,,methanol
Formula Weight 291.97
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 4'-Dibromopropiophenone
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger