Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-1-(4-nitrophenoxy)benzene, 98%, Thermo Scientific Chemicals

Catalog Number AAA1997714
Click to view available options
Quantity:
25 g
1836-75-5
C12H7Cl2NO3
284.092
MFCD00128026
XITQUSLLOSKDTB-UHFFFAOYSA-N
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
15787
2,4-dichloro-1-(4-nitrophenoxy)benzene
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
1836-75-5
C12H7Cl2NO3
284.092
MFCD00128026
XITQUSLLOSKDTB-UHFFFAOYSA-N
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
15787
2,4-dichloro-1-(4-nitrophenoxy)benzene
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 71°C
Quantity 25 g
UN Number UN3077
Beilstein 1887356
Merck Index 14,6598
Formula Weight 284.1
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-1-(4-nitrophenoxy)benzene
Hazard Category H302-H350-H360D
Hazard Statement GHS H Statement
H350-H360-H302
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 217-406-0
RTECSNumber KN8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit