Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-1-nitrobenzene, 97%

Manufacturer:  Alfa Aesar™ B237490B

Quantity 1000g

 View more versions of this product

Catalog No. AAB237490B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

611-06-3
192
UN2811
4-Nitro-m-dichlorobenzene
C6H3Cl2NO2
MFCD00007071
1451655
2,4-Dichloro-1-nitrobenzene

Specifications

1000g
C6H3Cl2NO2
MFCD00007071
258° to 259°C
29° to 33°C
4-Nitro-m-dichlorobenzene
97%
611-06-3
192
UN2811
1451655
152°C (306°F)
2,4-Dichloro-1-nitrobenzene

Safety and Handling

GHS H Statement
H311-H302-H317
Toxic in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P280h-P262-P273-P309-P310
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : CZ5420000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit