Alfa Aesar™ 2,4-Dichloro-1-nitrobenzene, 97%

Manufacturer: Alfa Aesar™ B2374930

Quantity 250g

 View more versions of this product

Catalog No. AAB2374930

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 611-06-3
    Molecular Formula C6H3Cl2NO2
    Formula Weight 192
    MDL Number MFCD00007071
    UN Number UN2811
    Beilstein 1451655
    Synonym 4-Nitro-m-dichlorobenzene
    Chemical Name or Material 2,4-Dichloro-1-nitrobenzene

    Specifications

    Formula Weight 192
    Boiling Point 258° to 259°C
    Flash Point 152°C (306°F)
    TSCA TSCA
    Molecular Formula C6H3Cl2NO2
    MDL Number MFCD00007071
    CAS 611-06-3
    UN Number UN2811
    Beilstein 1451655
    Melting Point 29° to 33°C
    Synonym 4-Nitro-m-dichlorobenzene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H317
    Toxic in contact with skin.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P280h-P262-P273-P309-P310
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number CZ5420000
    Quantity 250g
    Chemical Name or Material 2,4-Dichloro-1-nitrobenzene
    Assay 97%

    GHS H Statement
    H311-H302-H317
    Toxic in contact with skin.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P280h-P262-P273-P309-P310
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit