Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-3-ethyl-6-nitrophenol, 98%

Click to view available options
Quantity:
2.5g
10g
99817-36-4
C8H7Cl2NO3
236.048
MFCD00270764
YTVCECQSAPGJBB-UHFFFAOYSA-N
2,4-dichloro-3-ethyl-6-nitrophenol,2,4-dichloro-3-ethyl-6-nitro-phenol,phenol, 2,4-dichloro-3-ethyl-6-nitro,2-nitro-4,6-dichloro-5-ethylphenol,2,4-dichloro-3-ethyl-6-nitrochloride,pubchem20442,acmc-209se5,ksc486m4b,2,4-bis chloranyl-3-ethyl-6-nitro-phenol
7020339
CCC1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl
99817-36-4
C8H7Cl2NO3
236.048
MFCD00270764
YTVCECQSAPGJBB-UHFFFAOYSA-N
2,4-dichloro-3-ethyl-6-nitrophenol,2,4-dichloro-3-ethyl-6-nitro-phenol,phenol, 2,4-dichloro-3-ethyl-6-nitro,2-nitro-4,6-dichloro-5-ethylphenol,2,4-dichloro-3-ethyl-6-nitrochloride,pubchem20442,acmc-209se5,ksc486m4b,2,4-bis chloranyl-3-ethyl-6-nitro-phenol
7020339
CCC1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl
Melting Point 49°C to 51°C
Quantity 2.5g
UN Number UN2811
Formula Weight 236.05
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280-P301+P310-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c

H301-H317-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 420-740-1

TSCA : Yes

Recommended Storage : Ambient temperatures