Alfa Aesar™ 2,4-Dichloro-3-ethyl-6-nitrophenol, 98%

Manufacturer: Alfa Aesar™ B2226109

Quantity 10g

 View more versions of this product

Catalog No. AAB2226109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99817-36-4
    Molecular Formula C8H7Cl2NO3
    Formula Weight 236.05
    MDL Number MFCD00270764
    UN Number UN2811
    Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol

    Specifications

    Formula Weight 236.05
    TSCA TSCA
    Molecular Formula C8H7Cl2NO3
    MDL Number MFCD00270764
    CAS 99817-36-4
    UN Number UN2811
    Melting Point 49° to 51°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H318-H317
    Toxic if swallowed.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 10g
    Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol
    Assay 98%

    GHS H Statement
    H301-H318-H317
    Toxic if swallowed.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit