Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dichloro-5-fluorobenzamide, 97+%

Click to view available options
Quantity:
1g
5g
135748-33-3
C7H4Cl2FNO
208.013
MFCD01631478
SKSVSZRFINHARI-UHFFFAOYSA-N
acmc-20a3pc,2,4-dichloro-5-fluoro-benzamide,benzamide, 2,4-dichloro-5-fluoro,2,4-dichloro-5-fluorobenzamide
2774011
2,4-dichloro-5-fluorobenzamide
C1=C(C(=CC(=C1F)Cl)Cl)C(=O)N
135748-33-3
C7H4Cl2FNO
208.013
MFCD01631478
SKSVSZRFINHARI-UHFFFAOYSA-N
acmc-20a3pc,2,4-dichloro-5-fluoro-benzamide,benzamide, 2,4-dichloro-5-fluoro,2,4-dichloro-5-fluorobenzamide
2774011
2,4-dichloro-5-fluorobenzamide
C1=C(C(=CC(=C1F)Cl)Cl)C(=O)N
Quantity 1g
Melting Point 158°C to 160°C
Formula Weight 208.02
Percent Purity ≥97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning